Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025287.2 | NP_079563.2 | 4 | 280 | ||
GenBank mRNA | AB071989.1 | - | 2 | 89 | ||
AK004669.1 | - | 4 | 250 | |||
AK028201.1 | - | 3 | 416 | |||
AK030746.1 | - | 3 | 467 | |||
AK132383.1 | - | 3 | 456 | |||
AK159482.1 | - | 3 | 433 | |||
AK160858.1 | - | 3 | 377 | |||
AK166581.1 | - | 3 | 390 | |||
AK168185.1 | - | 3 | 402 | |||
AK168546.1 | - | 3 | 469 | |||
AK216748.1 | - | 2 | 64 | |||
AY538613.1 | - | 2 | 92 | |||
BC043131.1 | - | 3 | 299 | 6811159 | ||
BC045205.1 | - | 4 | 250 | 4988521 | ||
BC064809.1 | - | 3 | 404 | |||
BY303667.1 | - | 4 | 286 | |||
BY726358.1 | - | 4 | 281 |