Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009371.1 | - | 2 | 656 | ||
AK010055.1 | - | 2 | 656 | |||
AK010107.1 | - | 2 | 656 | |||
BC031537.1 | - | 2 | 645 |