Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167750.1 | NP_001161222.1 | 2, 3 | 243 | ||
NM_001167751.1 | NP_001161223.1 | 2, 3 | 243 | |||
NM_024260.5 | NP_077222.4 | 2, 3 | 243 | |||
GenBank mRNA | AK018415.1 | - | 2, 3 | 212 | ||
AK042031.1 | - | 2, 3 | 223 | |||
AK075898.1 | - | 2, 3 | 232 | |||
AK129458.1 | - | 2, 3 | 193 | |||
AY254697.1 | - | 2, 3 | 236 | |||
BC036294.1 | - | 2, 3 | 183 | 3500316 | ||
BC043092.1 | - | 2, 3 | 200 | 6405356 | ||
BY100563.1 | - | 2, 3 | 246 |