Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033865.1 | NP_001029037.1 | 3, 4 | 509 | ||
NM_024277.2 | NP_077239.1 | 4, 5 | 414 | |||
GenBank mRNA | AK018706.1 | - | 4, 5 | 236 | ||
BC002108.1 | - | 3, 4 | 224 | 3487291 | ||
BC081446.1 | - | 3, 4 | 215 | 6408051 | ||
BC096392.1 | - | 3, 4 | 228 | 1380923 |