Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024287.4 | NP_077249.1 | 3, 4 | 820 | ||
GenBank mRNA | AB041575.1 | - | 3, 4 | 619 | ||
AB232596.1 | - | 3, 4 | 182 | |||
AK051246.1 | - | 3, 4 | 655 | |||
AK083262.1 | - | 3, 4 | 644 | |||
AK084131.1 | - | 3, 4 | 628 | |||
AK160134.1 | - | 3, 4 | 699 | |||
AK160866.1 | - | 3, 4 | 628 | |||
AK169842.1 | - | 3, 4 | 821 | |||
AK183477.1 | - | 2, 3 | 103 | |||
AK185691.1 | - | 2, 3 | 103 | |||
BC026915.1 | - | 2, 3 | 110 |