Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Derl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024440.2 | NP_077760.1 | 5 | 454 | ||
GenBank mRNA | AB047555.1 | - | 5 | 454 | ||
AK007948.1 | - | 5 | 440 | |||
AK172429.1 | - | 5 | 440 | |||
BC004729.1 | - | 5 | 437 | |||
BC019815.1 | - | 5 | 434 | |||
BC099615.1 | - | 5 | 434 | |||
BC115466.1 | - | 5 | 449 | 40100387 | ||
BC115467.1 | - | 5 | 449 | 40100388 |