Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC005537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024473.3 | NP_077793.1 | 2 | 1894 | ||
GenBank mRNA | AK035718.1 | - | 2 | 1894 | ||
AK050530.1 | - | 689 | ||||
AK078765.1 | - | 2 | 1560 | |||
AK160094.1 | - | 851 | ||||
BC002217.1 | - | 268 | 3490515 | |||
BC005537.1 | - | 1531 |