Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synj2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025292.6 | NP_079568.1 | 2 | 319 | ||
GenBank mRNA | AK008054.1 | - | 2 | 302 | ||
AK008254.1 | - | 2 | 289 | |||
AK013474.1 | - | 2 | 281 | |||
AK033514.1 | - | 2 | 320 | |||
AK163362.1 | - | 2 | 287 | |||
AY071903.1 | - | 1 | 592 | |||
AY138960.1 | - | 2 | 259 | |||
AY566156.1 | - | 2 | 292 | |||
BC027433.1 | - | 2 | 281 |