Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024263.4 | NP_077225.4 | 10 | 1827 | ||
GenBank mRNA | AB040488.1 | - | 10 | 1762 | ||
AB052097.1 | - | 10 | 1720 | |||
AK004732.1 | - | 10 | 1710 | |||
BC002140.1 | - | 5 | 839 | |||
BC026438.1 | - | 10 | 1764 | 4239626 |