Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nts | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024435.2 | NP_077755.1 | 4 | 508 | ||
GenBank mRNA | AF304160.1 | - | 4 | 511 | ||
AK017202.1 | - | 4 | 526 | |||
AK017212.1 | - | 4 | 524 | |||
BC043024.1 | - | 4 | 508 | 5691298 |