Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ost4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134692.2 | NP_001128164.1 | 2 | 438 | ||
NM_024460.4 | NP_077780.3 | 3 | 310 | |||
GenBank mRNA | AA563474.1 | - | 3 | 310 | ||
AK002224.2 | - | 3 | 285 | |||
AK002453.1 | - | 3 | 232 | |||
AK003569.1 | - | 3 | 289 | |||
AK005413.1 | - | 3 | 233 | |||
AK009384.1 | - | 245 | ||||
AK010297.1 | - | 2 | 374 | |||
AK019285.1 | - | 3 | 270 | |||
BC002177.1 | - | 2 | 375 | 3486998 | ||
BC082316.1 | - | 2 | 349 | 6814413 | ||
CA466367.1 | - | 3 | 268 | |||
DV041491.1 | - | 351 |