Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024499.1 | NP_078775.1 | 12 | 1340 | ||
GenBank mRNA | AK039966.1 | - | 11 | 1335 | ||
AK051005.1 | - | 12 | 1339 | |||
AK079168.1 | - | 12 | 1350 | |||
AK165557.1 | - | 12 | 1364 | |||
AK183506.1 | - | 1 | 301 | |||
AK194124.1 | - | 1 | 24 | |||
BC003836.1 | - | 12 | 1340 | 3486148 |