Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025277.3 | NP_079553.1 | 3 | 572 | ||
GenBank mRNA | AK003187.1 | - | 3 | 510 | ||
AK014158.1 | - | 3 | 532 | |||
BC055434.1 | - | 4 | 497 | 3994982 | ||
BC061090.1 | - | 3 | 495 |