Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025281.3 | NP_079557.2 | 6 | 751 | ||
GenBank mRNA | AK006373.1 | - | 6 | 685 | ||
BC116923.1 | - | 5 | 607 | 40126242 | ||
BC116927.1 | - | 5 | 607 | 40126246 | ||
S60885.1 | - | 6 | 698 |