Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025290.3 | NP_079566.1 | 8, 9 | 970 | ||
GenBank mRNA | AB006535.1 | - | 8, 9 | 957 | ||
AK005739.1 | - | 8, 9 | 922 | |||
BC049584.1 | - | 8, 9 | 970 | 6744132 | ||
DQ141549.1 | - | 8, 9 | 869 | |||
DQ141550.1 | - | 8, 9 | 869 | |||
DQ141551.1 | - | 8, 9 | 869 | |||
DQ141554.1 | - | 8, 9 | 869 | |||
DQ141555.1 | - | 7, 8 | 755 |