Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025315.3 | NP_079591.1 | 3, 4 | 298 | ||
GenBank mRNA | AK009742.1 | - | 3, 4 | 299 | ||
AK012217.1 | - | 3, 4 | 299 | |||
AK014428.1 | - | 3, 4 | 292 | |||
AK019417.1 | - | 3, 4 | 288 | |||
BC012286.1 | - | 3, 4 | 289 | 3493056 | ||
BC096641.1 | - | 2, 3 | 1248 |