Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000089.3 | NP_000080.2 | 2, 4 | 560 | ||
GenBank mRNA | AK226074.1 | - | 2, 4 | 211 | ||
AK297786.1 | - | 2, 4 | 227 | |||
AK300194.1 | - | 2, 4 | 226 | |||
BC042586.1 | - | 2, 4 | 207 | 4803351 | ||
BC054498.1 | - | 2, 4 | 189 | 6474549 | ||
J03464.1 | - | 2, 4 | 558 | |||
KU177988.1 | - | 2, 4 | 89 | |||
KU177989.1 | - | 2, 4 | 89 | |||
Y00724.1 | - | 2, 4 | 165 | |||
Z74616.1 | - | 2, 4 | 228 |