Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025334.3 | NP_079610.1 | 1, 2 | 186 | ||
GenBank mRNA | AK002862.1 | - | 1, 2 | 173 | ||
AK003481.1 | - | 1, 2 | 167 | |||
AY548113.1 | - | 1, 2 | 98 | |||
BC006857.1 | - | 1, 2 | 186 | 2615934 |