Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110001J03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025363.3 | NP_079639.1 | 2 | 263 | ||
GenBank mRNA | AK003231.2 | - | 2 | 284 | ||
AK008291.2 | - | 2 | 264 | |||
AK009186.1 | - | 2 | 257 | |||
BC059710.1 | - | 2 | 279 | 6539235 |