Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifitm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025378.2 | NP_079654.1 | 1 | 179 | ||
GenBank mRNA | AK003407.1 | - | 1 | 180 | ||
AK007916.1 | - | 1 | 172 | |||
AK007919.1 | - | 1 | 168 | |||
AK151890.1 | - | 1 | 145 | |||
AY082484.1 | - | 1 | 148 | |||
AY594690.1 | - | 1 | 195 | |||
BC010291.1 | - | 1 | 131 | 3152819 |