Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025381.2 | NP_079657.1 | 1 | 165 | ||
GenBank mRNA | AK003419.1 | - | 1 | 162 | ||
AK007327.1 | - | 1 | 166 | |||
AK152153.1 | - | 1 | 162 | |||
AK152929.1 | - | 1 | 189 | |||
AK152959.1 | - | 1 | 162 | |||
BC016553.1 | - | 1 | 175 | 2812592 |