Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025412.2 | NP_079688.2 | 1, 2 | 193 | ||
GenBank mRNA | AK002912.1 | - | 1, 2 | 124 | ||
AK004291.1 | - | 1, 2 | 197 | |||
AK013214.1 | - | 1, 2 | 119 | |||
BC026536.1 | - | 1, 2 | 114 | 4987954 | ||
CX212991.1 | - | 1, 2 | 200 |