Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168470.1 | NP_001161942.1 | 2 | 579 | ||
NM_025318.3 | NP_079594.1 | 2 | 530 | |||
GenBank mRNA | AK002385.1 | - | 2 | 515 | ||
AK003198.1 | - | 2 | 525 | |||
AK004149.1 | - | 2 | 525 | |||
AK012769.1 | - | 2 | 515 | |||
BC022104.1 | - | 2 | 501 | 5101119 |