Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025332.3 | NP_079608.1 | 1, 2 | 373 | ||
GenBank mRNA | AK002771.1 | - | 1, 2 | 358 | ||
AK010979.1 | - | 1, 2 | 370 | |||
AK147840.1 | - | 1, 2 | 357 | |||
BC107216.1 | - | 1, 2 | 370 | 40051675 | ||
CJ169587.1 | - | 1, 2 | 374 |