Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025352.2 | NP_079628.1 | 2 | 365 | ||
GenBank mRNA | AA510422.1 | - | 2 | 310 | ||
AK002389.1 | - | 3 | 261 | |||
AK003160.1 | - | 3 | 277 | |||
AK005374.1 | - | 347 | ||||
AK007982.1 | - | 2 | 326 | |||
AK203034.1 | - | 2 | 113 | |||
AK214907.1 | - | 1 | 72 | |||
BC027809.1 | - | 1 | 1709 | |||
BC028519.1 | - | 2 | 349 | 1365708 | ||
CB950804.1 | - | 2 | 368 |