Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000108.4 | NP_000099.2 | 3 | 309 | ||
NM_001289751.1 | NP_001276680.1 | 3 | 309 | |||
NM_001289752.1 | NP_001276681.1 | 3 | 309 | |||
GenBank mRNA | AB209703.1 | - | 3 | 219 | ||
AK225289.1 | - | 3 | 233 | |||
AK225301.1 | - | 3 | 245 | |||
AK297508.1 | - | 3 | 243 | |||
AK300077.1 | - | 3 | 242 | |||
AK310220.1 | - | 3 | 230 | |||
AK312346.1 | - | 3 | 256 | |||
BC018648.2 | - | 3 | 218 | 3029752 | ||
BC018696.2 | - | 3 | 218 | 3029752 | ||
DC354209.1 | - | 3 | 309 | |||
J03490.1 | - | 3 | 241 | |||
J03620.1 | - | 3 | 215 |