Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPYD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000110.3 | NP_000101.2 | 5 | 586 | ||
NM_001160301.1 | NP_001153773.1 | 5 | 586 | |||
XM_005270562.3 | XP_005270619.2 | 5 | 586 | |||
XM_006710397.3 | XP_006710460.1 | 5 | 586 | |||
XM_017000507.1 | XP_016855996.1 | 4 | 469 | |||
XM_017000508.1 | XP_016855997.1 | 5 | 581 | |||
XM_017000509.1 | XP_016855998.1 | 5 | 581 | |||
XM_017000510.1 | XP_016855999.1 | 4 | 470 | |||
GenBank mRNA | AB003063.1 | - | 5 | 449 | ||
AK291217.1 | - | 5 | 552 | |||
AK297515.1 | - | 4 | 413 | |||
BC008379.1 | - | 5 | 473 | |||
BC064027.1 | - | 5 | 512 | |||
BC108742.1 | - | 5 | 545 | |||
BC131777.1 | - | 5 | 502 | 40123875 | ||
BC131778.1 | - | 5 | 502 | 40123878 | ||
BT006740.1 | - | 5 | 449 | |||
U09178.1 | - | 5 | 530 | |||
U20938.1 | - | 5 | 550 |