Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163525.2 | NP_001156997.1 | 4 | 535 | ||
NM_001163526.2 | NP_001156998.1 | 3 | 371 | |||
NM_025393.4 | NP_079669.2 | 4 | 518 | |||
GenBank mRNA | AK003669.1 | - | 4 | 498 | ||
AK019075.1 | - | 4 | 489 | |||
AK132637.1 | - | 2 | 1135 | |||
BC025607.1 | - | 3 | 517 | 5151924 | ||
W54064.1 | - | 2 | 335 |