Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025400.3 | NP_079676.1 | 6 | 641 | ||
GenBank mRNA | AK003948.1 | - | 6 | 595 | ||
AK009262.1 | - | 5 | 371 | |||
AK020083.1 | - | 6 | 624 | |||
AK082469.1 | - | 6 | 581 | |||
AK148048.1 | - | 6 | 673 | |||
AK154593.1 | - | 5 | 772 | |||
BC011315.1 | - | 6 | 641 | 4013700 |