Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025407.2 | NP_079683.2 | 3 | 347 | ||
GenBank mRNA | AK002292.1 | - | 3 | 288 | ||
AK004031.1 | - | 3 | 293 | |||
AK010553.1 | - | 3 | 287 | |||
AK013128.1 | - | 3 | 287 | |||
AK151764.1 | - | 3 | 289 | |||
AK160337.1 | - | 3 | 294 | |||
AK167911.1 | - | 3 | 287 | |||
AK168218.1 | - | 3 | 289 | |||
AK177346.1 | - | 2 | 140 | |||
AK177461.1 | - | 2 | 140 | |||
AK184915.1 | - | 2 | 140 | |||
BB649589.1 | - | 3 | 347 | |||
BC030064.1 | - | 3 | 284 | 1400271 | ||
BY260300.1 | - | 3 | 320 |