Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ftsj3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025310.2 | NP_079586.1 | 21 | 2563 | ||
GenBank mRNA | AK004996.2 | - | 21 | 2563 | ||
AK031514.1 | - | 21 | 2550 | |||
AK145490.1 | - | 21 | 2588 | |||
BC012281.1 | - | 21 | 2560 | 4012961 |