Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem254a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025311.3 | NP_079587.1 | 926 | |||
NM_026679.2 | NP_080955.1 | 1095 | ||||
GenBank mRNA | AK002342.1 | - | 4 | 914 | ||
AK002441.1 | - | 4 | 1026 | |||
AK003554.1 | - | 4 | 835 | |||
AK008299.1 | - | 4 | 886 | |||
BC031854.1 | - | 4 | 979 | 4194400 |
Gene Symbol: Tmem254b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270495.1 | NP_001257424.1 | 4 | 1079 | ||
NM_001270496.1 | NP_001257425.1 | 4 | 837 | |||
GenBank mRNA | DV046977.1 | - | 3 | 697 |
Gene Symbol: Tmem254c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270498.1 | NP_001257427.1 | 1095 | |||
NM_001270499.1 | NP_001257428.1 | 915 | ||||
GenBank mRNA | BY444156.1 | - | 66 | |||
BY703810.1 | - | 835 |