Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdhaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025333.4 | NP_079609.2 | 4 | 1055 | ||
GenBank mRNA | AK013454.1 | - | 4 | 960 | ||
AK075594.1 | - | 4 | 960 | |||
AK160175.1 | - | 2 | 688 | |||
AK160957.1 | - | 4 | 962 | |||
AK167228.1 | - | 4 | 991 | |||
BC029630.1 | - | 3 | 774 | |||
BC038924.1 | - | 3 | 1023 | |||
BC048913.1 | - | 4 | 788 | |||
BC085277.1 | - | 4 | 957 | 30441476 | ||
BU695785.1 | - | 1 | 165 |