Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025404.3 | NP_079680.1 | 2 | 1138 | ||
GenBank mRNA | AK004126.1 | - | 2 | 1119 | ||
AK013320.1 | - | 2 | 1116 | |||
BC016113.1 | - | 2 | 1104 | 4481779 |