Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025414.3 | NP_079690.2 | 24 | 3160 | ||
GenBank mRNA | AK018609.1 | - | 9 | 1190 | ||
AK080117.1 | - | 22 | 2879 | |||
BC007156.1 | - | 17 | 2076 | 3486558 | ||
BC046294.1 | - | 23 | 2883 | |||
BC060115.1 | - | 21 | 4605 |