Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025430.3 | NP_079706.1 | 2 | 132 | ||
GenBank mRNA | AK004385.1 | - | 2 | 125 | ||
AK010223.1 | - | 2 | 121 | |||
AK010369.1 | - | 2 | 118 | |||
AK012807.1 | - | 2 | 119 | |||
AK039187.1 | - | 2 | 112 | |||
BB615775.1 | - | 2 | 132 | |||
BC028750.1 | - | 2 | 124 | 1245736 |