Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lbhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286103.1 | NP_001273032.1 | 4 | 618 | ||
GenBank mRNA | CK619869.1 | - | 4 | 588 |
Gene Symbol: 1810009A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025463.3 | NP_079739.1 | 2 | 122 | ||
GenBank mRNA | AK007390.1 | - | 2 | 103 | ||
AK012061.1 | - | 2 | 112 | |||
BC047099.1 | - | 2 | 101 | 5253822 | ||
BC058702.1 | - | 2 | 85 | 6822952 | ||
BY264380.1 | - | 2 | 124 | |||
BY273399.1 | - | 1 | 265 |