Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810032O08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007675.1 | - | 3 | 317 | ||
AK013681.1 | - | 2 | 116 | |||
AK037979.1 | - | 3 | 308 | |||
AK088915.1 | - | 3 | 299 | |||
BC055707.1 | - | 3 | 302 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_025472.1 | - | 308 | Chr. - - [-] on Build |