Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025478.3 | NP_079754.2 | 2 | 389 | ||
GenBank mRNA | AK011674.1 | - | 2 | 256 | ||
AK134794.1 | - | 2 | 389 | |||
AK152763.1 | - | 2 | 386 | |||
AK153257.1 | - | 2 | 386 | |||
AK165542.1 | - | 2 | 408 | |||
AY035213.1 | - | 2 | 389 | |||
BC016576.1 | - | 2 | 396 | 2609919 |