Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291197.1 | NP_001278126.1 | 2, 3 | 345 | ||
NM_001291200.1 | NP_001278129.1 | 2, 3 | 345 | |||
NM_001291201.1 | NP_001278130.1 | 2, 3 | 345 | |||
NM_001291202.1 | NP_001278131.1 | 2, 3 | 345 | |||
NM_001291203.1 | NP_001278132.1 | 2, 3 | 345 | |||
NM_025482.3 | NP_079758.2 | 2, 3 | 345 | |||
GenBank mRNA | AK013085.1 | - | 2, 3 | 287 | ||
AK013188.1 | - | 2, 3 | 298 | |||
AK051268.1 | - | 2, 3 | 289 | |||
AK147885.1 | - | 2, 3 | 305 | |||
AK155987.1 | - | 2, 3 | 282 | |||
AK159925.1 | - | 2, 3 | 273 | |||
AK160727.1 | - | 2, 3 | 289 | |||
AK161092.1 | - | 2, 3 | 301 | |||
AK171825.1 | - | 2, 3 | 307 | |||
BC006886.1 | - | 2, 3 | 234 | 2647702 | ||
CN721218.1 | - | 1, 2 | 199 | |||
DV047279.1 | - | 2, 3 | 331 |