Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scp2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025490.2 | NP_079766.3 | 1 | 190 | ||
GenBank mRNA | AK005847.1 | - | 1 | 190 | ||
BC048653.1 | - | 1 | 190 | 6773017 |