Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025491.3 | NP_079767.2 | 4 | 537 | ||
NM_028340.1 | NP_082616.1 | 4 | 706 | |||
GenBank mRNA | AK003860.1 | - | 4 | 537 | ||
AK006068.1 | - | 4 | 527 | |||
BC045525.1 | - | 4 | 706 | 6468669 |