Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000132.3 | NP_000123.1 | 24 | 6876 | ||
NM_019863.2 | NP_063916.1 | 3 | 445 | |||
GenBank mRNA | AK289947.1 | - | 3 | 375 | ||
AK295164.1 | - | 12 | 1673 | |||
AK313707.1 | - | 3 | 330 | |||
AX746360.1 | - | 24 | 6705 | |||
BC022513.1 | - | 3 | 364 | |||
BC064380.1 | - | 3 | 424 | |||
BC098389.1 | - | 3 | 443 | |||
BC111967.1 | - | 3 | 333 | |||
BC111969.1 | - | 3 | 332 | |||
BI547023.1 | - | 3 | 484 | |||
K01740.1 | - | 24 | 6855 | |||
KT878539.1 | - | 1 | 126 | |||
M14113.1 | - | 24 | 6876 | |||
M90707.1 | - | 3 | 340 | |||
X01179.1 | - | 24 | 6814 |