Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025519.2 | NP_079795.1 | 2 | 268 | ||
GenBank mRNA | AK008217.1 | - | 1 | 1184 | ||
AK009273.1 | - | 2 | 268 | |||
AK010707.1 | - | 2 | 335 | |||
AK145033.1 | - | 2 | 272 | |||
AK160189.1 | - | 2 | 262 | |||
AK166371.1 | - | 2 | 290 | |||
BC115983.1 | - | 2 | 258 | 40105850 | ||
BC117538.1 | - | 2 | 258 | 40105849 |