Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025529.3 | NP_079805.1 | 3 | 415 | ||
GenBank mRNA | AK002605.1 | - | 3 | 353 | ||
AK009700.1 | - | 3 | 353 | |||
AK045197.1 | - | 3 | 416 | |||
AK170076.1 | - | 3 | 355 | |||
BC056443.1 | - | 3 | 355 | 6822891 |