Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025444.2 | NP_079720.1 | 4 | 266 | ||
GenBank mRNA | AK007227.1 | - | 4 | 272 | ||
AK007339.1 | - | 4 | 267 | |||
AK076595.1 | - | 266 | ||||
AK160171.1 | - | 4 | 266 | |||
BC021447.1 | - | 4 | 278 | 5050736 |