Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dimt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025447.4 | NP_079723.1 | 2, 3 | 237 | ||
GenBank mRNA | AK007759.1 | - | 2, 3 | 188 | ||
AK011544.1 | - | 2, 3 | 194 | |||
AK015145.1 | - | 2, 3 | 187 | |||
AK078186.1 | - | 2, 3 | 189 | |||
AK078401.1 | - | 2, 3 | 211 | |||
AK146188.1 | - | 2, 3 | 208 | |||
AK161333.1 | - | 2, 3 | 189 | |||
BC019799.1 | - | 2, 3 | 210 | 5053500 | ||
BF608861.1 | - | 2, 3 | 248 |