Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025454.2 | NP_079730.1 | 4 | 382 | ||
GenBank mRNA | AK006421.1 | - | 4 | 481 | ||
AK007536.1 | - | 4 | 373 | |||
AK009312.1 | - | 4 | 366 | |||
AK028010.1 | - | 4 | 360 | |||
AK028397.1 | - | 4 | 383 | |||
AK145695.1 | - | 4 | 355 | |||
BC064674.1 | - | 4 | 381 | 6400183 |