Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025461.5 | NP_079737.1 | 4 | 366 | ||
GenBank mRNA | AK007450.1 | - | 4 | 343 | ||
AK007562.1 | - | 4 | 355 | |||
AK008486.1 | - | 4 | 331 | |||
AK040003.1 | - | 4 | 368 | |||
AK082374.1 | - | 4 | 367 |