Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810009A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025463.3 | NP_079739.1 | 4 | 387 | ||
GenBank mRNA | AK007390.1 | - | 4 | 368 | ||
BC047099.1 | - | 4 | 366 | 5253822 | ||
BC058702.1 | - | 4 | 350 | 6822952 | ||
BY264380.1 | - | 4 | 390 |
Gene Symbol: Lbhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286103.1 | NP_001273032.1 | 6 | 883 |